This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00499987_m1
See other CDHR1 GE Assays ›
Gene Symbol:
Cdhr1
Gene Name:
cadherin-related family member 1
Gene Aliases:
Pcdh21, Prcad, mKIAA1775
Chromosome Location:
Chr.14: 37077849 - 37098311 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00499987_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdhr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_130878.2 NP_570948.1 9 - 10 1031 94
GenBank mRNA AB093300.1 - 9 - 10 1447 94
AF426393.1 - 9 - 10 918 94
AK143151.1 - 9 - 10 972 94
BC042454.1 - 9 - 10 941 5369576 94
BC058270.1 - 9 - 10 1031 6490597 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001745.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdhr1

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
cellular process
photoreceptor cell maintenance
calcium ion binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400013585.mm.1 NM_130878.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400013585.mm.1 NM_130878.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400013585.mm.1 TC1400001745.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0