This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00500458_g1
See other IL15RA GE Assays ›
Gene Symbol:
Il15ra
Gene Name:
interleukin 15 receptor, alpha chain
Gene Aliases:
AA690181, IL-15RA
Chromosome Location:
Chr.2: 11705169 - 11733985 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00500458_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il15ra
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271497.1 NP_001258426.1 4 - 5 653 83
NM_001271499.1 NP_001258428.1 4 - 5 617 83
NM_008358.2 NP_032384.1 4 - 5 716 83
NM_133836.2 NP_598597.1 4 - 5 769 83
GenBank mRNA AY219715.1 - 4 - 5 518 83
AY221616.1 - 4 - 5 394 83
BC022705.1 - 4 - 5 526 83
BC095982.1 - 1 - 2 444 83
BC132231.1 - 3 - 4 316 83
BC132233.1 - 3 - 4 316 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000222.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Il15ra

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s)

JAK-STAT cascade
negative regulation of neuron projection development
positive regulation of natural killer cell differentiation

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline