This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00500822_g1
See other NQO1 GE Assays ›
Gene Symbol:
Nqo1
Gene Name:
NAD(P)H dehydrogenase, quinone 1
Gene Aliases:
AV001255, Dia4, Dtd, Nmo-1, Nmo1, Nmor1, Ox-1, Ox1, Qr1
Chromosome Location:
Chr.8: 107388225 - 107403205 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00500822_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nqo1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008706.5 NP_032732.3 2 - 3 307 88
GenBank mRNA AK075568.1 - 2 - 3 308 88
BC004579.1 - 2 - 3 285 3586888 88
CT010284.1 - 1 - 2 174 88
S75951.1 - 2 - 3 381 88
U12961.1 - 2 - 3 282 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002877.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nqo1

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

superoxide metabolic process
response to oxidative stress
removal of superoxide radicals
negative regulation of catalytic activity
positive regulation of neuron apoptotic process
oxidation-reduction process
NAD(P)H dehydrogenase (quinone) activity
superoxide dismutase activity
oxidoreductase activity
identical protein binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z47m5:80/100.66.77.18:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0