This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00501038_m1
See other PDE9A GE Assays ›
Gene Symbol:
Pde9a
Gene Name:
phosphodiesterase 9A
Gene Aliases:
PDE9A1
Chromosome Location:
Chr.17: 31386212 - 31476309 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00501038_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde9a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163748.1 NP_001157220.1 2 - 3 195 73
NM_008804.4 NP_032830.3 2 - 3 195 73
GenBank mRNA AF031147.1 - 2 - 3 150 73
AF068247.1 - 2 - 3 159 73
AK030509.1 - 2 - 3 179 73
AK168143.1 - 2 - 3 177 73
BC025882.1 - 2 - 3 153 73
BC038675.1 - 2 - 3 144 73
BC046915.1 - 2 - 3 144 73
BC061163.1 - 2 - 3 175 6533665 73
BY283127.1 - 2 - 3 196 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000535.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde9a

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
3',5'-cyclic-nucleotide phosphodiesterase activity
phosphoric diester hydrolase activity
hydrolase activity
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700004889.mm.1 NM_001163748.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700004889.mm.1 NM_001163748.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700004889.mm.1 NM_008804.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700004889.mm.1 NM_008804.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700004889.mm.1 TC1700000535.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline