This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00501210_m1
See other PTPN14 GE Assays ›
Gene Symbol:
Ptpn14
Gene Name:
protein tyrosine phosphatase, non-receptor type 14
Gene Aliases:
C130080N23Rik, OTTMUSG00000022087, PTP36
Chromosome Location:
Chr.1: 189728216 - 189876695 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00501210_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptpn14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008976.2 NP_033002.2 7 - 8 1202 62
GenBank mRNA AF170902.1 - 6 - 7 769 62
AF170903.1 - 6 - 7 769 62
AK028783.1 - 7 - 8 1166 62
AK037646.1 - 7 - 8 1203 62
AK086524.1 - 7 - 8 1196 62
AK156480.1 - 7 - 8 948 62
D31842.1 - 6 - 7 799 62

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptpn14

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

lymphangiogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
protein dephosphorylation
negative regulation of cell proliferation
dephosphorylation
peptidyl-tyrosine dephosphorylation
regulation of protein export from nucleus
regulation of nucleic acid-templated transcription
transcription cofactor activity
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
hydrolase activity
phosphatase activity
receptor tyrosine kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0