This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00501244_m1
See other PTPN22 GE Assays ›
Gene Symbol:
Ptpn22
Gene Name:
protein tyrosine phosphatase, non-receptor type 22 (lymphoid)
Gene Aliases:
70zpep, PEP, Ptpn8
Chromosome Location:
Chr.3: 103856804 - 103912252 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00501244_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptpn22
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008979.2 NP_033005.1 18 - 19 2355 102
NR_104070.1 20 - 21 2372 102
GenBank mRNA AK153812.1 - 20 - 21 2373 102
BC055377.1 - 18 - 19 2277 30077916 102
M90388.1 - 18 - 19 2340 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001064.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptpn22

Gene Ontology Categories:

Function(s) Process(es)

immune system process
protein dephosphorylation
autophagy
dephosphorylation
T cell differentiation
regulation of natural killer cell proliferation
peptidyl-tyrosine dephosphorylation
phosphoanandamide dephosphorylation
T cell receptor signaling pathway
regulation of T cell receptor signaling pathway
negative regulation of T cell receptor signaling pathway
negative regulation of T cell activation
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein binding
hydrolase activity
phosphatase activity
SH3 domain binding
kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0