This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00501291_g1
See other IGDCC3 GE Assays ›
Gene Symbol:
Igdcc3
Gene Name:
immunoglobulin superfamily, DCC subclass, member 3
Gene Aliases:
2810401C09Rik, AI851425, Punc, WI-14920
Chromosome Location:
Chr.9: 65141121 - 65187031 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00501291_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igdcc3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008988.2 NP_033014.1 4 - 5 943 90
GenBank mRNA AF026465.1 - 4 - 5 943 90
AK011916.1 - 4 - 5 979 90
AK051027.1 - 4 - 5 944 90
BC053057.1 - 4 - 5 920 5698028 90
BC086481.1 - 4 - 5 927 6505546 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000830.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igdcc3

Gene Ontology Categories:

Function(s) Process(es)

neuromuscular process controlling balance
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900006324.mm.1 NM_008988.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006324.mm.1 NM_008988.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900006324.mm.1 TC0900000830.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900006323.mm.1 NM_008988.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006323.mm.1 NM_008988.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900006323.mm.1 TC0900000830.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0