This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00501506_m1
See other TCF7L2 GE Assays ›
Gene Symbol:
Tcf7l2
Gene Name:
transcription factor 7 like 2, T cell specific, HMG box
Gene Aliases:
TCF4B, TCF4E, Tcf-4, Tcf4, mTcf-4B, mTcf-4E
Chromosome Location:
Chr.19: 55741810 - 55933661 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00501506_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tcf7l2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001142918.1 NP_001136390.1 6 - 7 1253 82
NM_001142919.1 NP_001136391.1 6 - 7 1253 82
NM_001142921.1 NP_001136393.1 6 - 7 1253 82
NM_001142922.1 NP_001136394.1 6 - 7 1253 82
NM_001142923.1 NP_001136395.1 6 - 7 1253 82
NM_001142924.1 NP_001136396.1 3 - 4 555 82
NM_009333.3 NP_033359.2 6 - 7 1253 82
GenBank mRNA AF107298.1 - 6 - 7 997 82
AF363724.1 - 7 - 8 953 82
AF363725.1 - 6 - 7 811 82
AJ223070.1 - 6 - 7 731 82
AK038720.1 - 6 - 7 699 82
AK048536.1 - 3 - 4 368 82
AY072035.1 - 6 - 7 794 82
BC009807.1 - 6 - 7 758 82
BC011397.1 - 6 - 7 1232 82
BC052022.1 - 6 - 7 832 5702449 82
CA750374.1 - 3 - 4 555 82

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tcf7l2

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
blood vessel development
cellular glucose homeostasis
glycogen metabolic process
glucose metabolic process
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
cell cycle arrest
cell proliferation
cellular response to starvation
response to glucose
post-embryonic development
positive regulation of triglyceride biosynthetic process
positive regulation of heparan sulfate proteoglycan biosynthetic process
oligodendrocyte development
Wnt signaling pathway
neural tube development
pituitary gland development
bone mineralization
negative regulation of BMP signaling pathway
embryonic genitalia morphogenesis
regulation of myelination
positive regulation of insulin secretion
positive regulation of protein binding
secretory granule localization
regulation of hormone metabolic process
somatic stem cell population maintenance
multicellular organism growth
catenin import into nucleus
regulation of gluconeogenesis by regulation of transcription from RNA polymerase II promoter
negative regulation of fibroblast growth factor receptor signaling pathway
odontogenesis of dentin-containing tooth
glucose homeostasis
positive regulation of apoptotic process
negative regulation of sequence-specific DNA binding transcription factor activity
maintenance of DNA repeat elements
skin development
canonical Wnt signaling pathway involved in positive regulation of epithelial to mesenchymal transition
fat cell differentiation
negative regulation of fat cell differentiation
positive regulation of gluconeogenesis
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of organ growth
positive regulation of protein export from nucleus
positive regulation of lipid biosynthetic process
embryonic digestive tract morphogenesis
embryonic hindgut morphogenesis
myoblast fate commitment
regulation of skeletal muscle tissue development
regulation of smooth muscle cell proliferation
generation of neurons
regulation of oligodendrocyte differentiation
positive regulation of epithelial cell proliferation
positive regulation of protein kinase B signaling
canonical Wnt signaling pathway
face morphogenesis
regulation of insulin secretion involved in cellular response to glucose stimulus
negative regulation of canonical Wnt signaling pathway
insulin metabolic process
negative regulation of type B pancreatic cell apoptotic process
negative regulation of extrinsic apoptotic signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II repressing transcription factor binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
beta-catenin binding
transcription factor binding
protein kinase binding
nuclear hormone receptor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
gamma-catenin binding
armadillo repeat domain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0