This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00501533_m1
See other AANAT GE Assays ›
Gene Symbol:
Aanat
Gene Name:
arylalkylamine N-acetyltransferase
Gene Aliases:
AA-NAT, Nat-2, Nat4, Snat
Chromosome Location:
Chr.11: 116591721 - 116597680 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00501533_m1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aanat
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009591.3 NP_033721.1 1 - 2 86 64
NR_033223.1 1 - 2 86 64
GenBank mRNA AF004108.1 - 1 - 2 86 64
AF004110.1 - 1 - 2 86 64
BC116967.1 - 1 - 2 57 40126286 64
BC119139.1 - 1 - 2 57 8733888 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001937.mm.1

Additional Information:

This assay is in the 5'UTR region of the AF004108.1,AF004110.1,BC116967.1,BC119139.1,NM_009591.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aanat

Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

N-terminal protein amino acid acetylation
circadian rhythm
response to zinc ion
response to organic cyclic compound
melatonin biosynthetic process
response to insulin
response to prostaglandin E
response to copper ion
rhythmic process
cellular response to cAMP
aralkylamine N-acetyltransferase activity
arylamine N-acetyltransferase activity
N-acetyltransferase activity
transferase activity
transferase activity, transferring acyl groups
14-3-3 protein binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158384 NM_009591.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158384 NR_033223.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158384 AF004109.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158384 AF004110.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158384 AF004111.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P497424 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481723 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233782 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887632 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 83162 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137605 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P497424 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158384 NM_009591.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158384 NR_033223.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158384 AF004109.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158384 AF004110.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158384 AF004111.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160256 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160247 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160261 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160259 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160255 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160272 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160263 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160252 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160274 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160258 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160267 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160257 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160270 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160249 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160260 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160254 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160265 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160269 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160251 - -
Whole Mouse Genome G4122A A_52_P497424 - -
Whole Mouse Genome G4122A A_51_P295511 NM_009591.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P295511 NR_033223.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P295511 AF004108.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P295511 AF004109.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P295511 AF004110.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131928 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09474817 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09372267 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09372275 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160240 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160238 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160233 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160241 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160276 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160212 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160284 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160257 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160236 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160273 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160263 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160274 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160287 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160278 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160268 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160228 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160256 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160261 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160252 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160235 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160255 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160223 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160231 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160227 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160272 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160258 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160254 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160285 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160281 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160259 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160279 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160251 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160217 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160243 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160247 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160215 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160260 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160226 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160247 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160238 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160281 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09702208 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09260703 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160241 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160254 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160287 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09372270 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160267 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160274 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160245 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160284 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160242 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160263 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160273 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160256 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160276 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09007243 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160259 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09114991 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09653851 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160233 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160261 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160257 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160236 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160260 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160285 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160258 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160252 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160235 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09372269 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160230 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09391849 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160268 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160272 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160278 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09702205 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28160251 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10382822 - -
Affymetrix Mouse Gene 1.0 ST Array 10382817 - -
Affymetrix Mouse Gene 1.0 ST Array 10382828 - -
Affymetrix Mouse Gene 1.0 ST Array 10382824 - -
Affymetrix Mouse Gene 1.0 ST Array 10382819 - -
Affymetrix Mouse Gene 1.0 ST Array 10382826 - -
Affymetrix Mouse Gene 1.0 ST Array 10382820 - -
Affymetrix Mouse Gene 1.0 ST Array 10382818 - -
Affymetrix Mouse Gene 1.0 ST Array 10382825 - -
Affymetrix Mouse Gene 1.0 ST Array 10382829 - -
Affymetrix Mouse Gene 1.0 ST Array 10382827 - -
Affymetrix Mouse Gene 1.0 ST Array 10382823 - -
Affymetrix HT MG-430 PM Array Plate 1421666_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10382816 - -
Mouse Genome 430 2.0 Array 1421666_a_at - -
Mouse Genome 430 2.0 Array 1421666_PM_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 11298_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 11298_at - -
Affymetrix Mouse Clariom S Array PSR1100018403.mm.1 NM_009591.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100018403.mm.1 NM_009591.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100018403.mm.1 NR_033223.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100018403.mm.1 NR_033223.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100018403.mm.1 TC1100001937.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100018404.mm.1 NM_009591.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100018404.mm.1 NM_009591.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100018404.mm.1 NR_033223.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100018404.mm.1 NR_033223.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100018404.mm.1 TC1100001937.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100018402.mm.1 NM_009591.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100018402.mm.1 NM_009591.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100018402.mm.1 NR_033223.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100018402.mm.1 NR_033223.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100018402.mm.1 TC1100001937.mm.1 MTA TC
Mouse Expression Set 430 1421666_a_at - -
Mouse Expression Set 430 1421666_PM_a_at - -
Murine Genome U74v2 Set 92511_at - -
Murine Genome U74v2 Set 92512_PM_g_at - -
Murine Genome U74v2 Set 92512_g_at - -
Murine Genome U74v2 Set 92511_PM_at - -
Mouse Genome 430A 2.0 Array 1421666_a_at - -
Mouse Genome 430A 2.0 Array 1421666_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1421666_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1421666_a_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 610609 - -
Illumina mouse-6 v1.0 expression beadchip 610609 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000610609 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2671661 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2671661 - -
mouse-6 v1.1 (Illumina) scl40703.5.1_48-S - -
Illumina Sentrix mouse-6.v1 bead array 610609 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl40703.5.1_48-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 610609 - -
MouseRef-8 v2 Expression BeadChip ILMN_2671661 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2671661 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0