This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00501826_g1
See other PPP2R3D GE Assays ›
Gene Symbol:
Ppp2r3d
Gene Name:
protein phosphatase 2 (formerly 2A), regulatory subunit B'', delta
Gene Aliases:
PP2A B''-PR59, PP2A PR59, PR59, Ppp2r3, Ppp2r3a
Chromosome Location:
Chr.9: 124438769 - 124441010 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00501826_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp2r3d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163415.1 NP_001156887.1 1640 91
NM_011154.2 NP_035284.2 1406 91
NR_102716.1 1897 91
GenBank mRNA AF050165.1 - 1403 91
AK143212.1 - 1683 91
BC027415.1 - 1383 4235453 91
BC059852.1 - 1671 6402777 91
BC096544.1 - 1086 6834351 91

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp2r3d

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
negative regulation of cell proliferation
regulation of protein phosphatase type 2A activity
negative regulation of G1/S transition of mitotic cell cycle
chromatin binding
protein binding
protein phosphatase type 2A regulator activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lzcpb:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0