This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00502050_g1
See other SLC7A10 GE Assays ›
Gene Symbol:
Slc7a10
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 10
Gene Aliases:
AL024237, Asc-1, D7Bwg0847e
Chromosome Location:
Chr.7: 35186357 - 35201114 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00502050_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017394.4 NP_059090.3 7 - 8 1133 90
GenBank mRNA AB026688.1 - 7 - 8 1047 90
AK005282.1 - 5 - 6 530 90
AK031867.1 - 7 - 8 1101 90
BC054765.1 - 7 - 8 1076 6414759 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000655.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc7a10

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
amino acid transport
neutral amino acid transport
L-serine transport
D-alanine transport
D-serine transport
cation transmembrane transport
L-alpha-amino acid transmembrane transport
protein binding
amino acid transmembrane transporter activity
neutral amino acid transmembrane transporter activity
L-serine transmembrane transporter activity
antiporter activity
D-serine transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700005503.mm.1 NM_017394.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700005503.mm.1 NM_017394.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700005503.mm.1 TC0700000655.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700005501.mm.1 NM_017394.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700005501.mm.1 NM_017394.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700005501.mm.1 TC0700000655.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline