This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00502437_m1
See other BCO1 GE Assays ›
Gene Symbol:
Bco1
Gene Name:
beta-carotene oxygenase 1
Gene Aliases:
Bcdo, Bcdo1, Bcmo1, CMO1, Cmoi, beta-CD, betaCMOOX
Chromosome Location:
Chr.8: 117095851 - 117133720 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00502437_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bco1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163028.1 NP_001156500.1 5 - 6 785 97
NM_021486.3 NP_067461.2 5 - 6 785 97
GenBank mRNA AF271298.1 - 5 - 6 816 97
AF294899.1 - 5 - 6 798 97
AJ278064.2 - 5 - 6 797 97
AK054171.1 - 4 - 5 537 97
AK146472.1 - 5 - 6 783 97
BC125328.1 - 5 - 6 652 40129843 97
BC125330.1 - 5 - 6 652 40129845 97
BC145036.1 - 5 - 6 652 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001396.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bco1

Panther Classification:

Molecular Function -

oxygenase

Gene Ontology Categories:

Function(s) Process(es)

retinoid metabolic process
oxidation-reduction process
beta-carotene 15,15'-monooxygenase activity
monooxygenase activity
oxidoreductase activity
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800011333.mm.1 NM_001163028.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011333.mm.1 NM_001163028.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011333.mm.1 NM_021486.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011333.mm.1 NM_021486.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800011333.mm.1 TC0800001396.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800011332.mm.1 NM_001163028.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011332.mm.1 NM_001163028.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011332.mm.1 NM_021486.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011332.mm.1 NM_021486.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800011332.mm.1 TC0800001396.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 644071 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0