This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00503205_g1
See other GET4 GE Assays ›
Gene Symbol:
Get4
Gene Name:
golgi to ER traffic protein 4 homolog (S. cerevisiae)
Gene Aliases:
1110007L15Rik, AW412535, Cee
Chromosome Location:
Chr.5: 139252324 - 139270050 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00503205_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Get4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163316.1 NP_001156788.1 3 - 4 264 78
NM_026269.2 NP_080545.2 3 - 4 455 78
GenBank mRNA AK003551.1 - 3 - 4 404 78
BC019557.1 - 3 - 4 246 4163641 78
CJ048159.1 - 3 - 4 263 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001669.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Get4

Gene Ontology Categories:

Function(s) Process(es)

transport
tail-anchored membrane protein insertion into ER membrane
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500015402.mm.1 TC0500001669.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500015402.mm.1 NM_001163316.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500015402.mm.1 NM_001163316.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500015402.mm.1 NM_026269.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500015402.mm.1 NM_026269.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0