This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00503428_m1
See other SYF2 GE Assays ›
Gene Symbol:
Syf2
Gene Name:
SYF2 homolog, RNA splicing factor (S. cerevisiae)
Gene Aliases:
1110018L13Rik, Cbpin, D4Bwg1551e, Gcipip, Ntc31, mp29, p29
Chromosome Location:
Chr.4: 134930980 - 134937537 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00503428_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Syf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026780.3 NP_081056.1 6 - 7 576 71
GenBank mRNA AK003787.1 - 6 - 7 578 71
AK152794.1 - 6 - 7 565 71
AY033432.1 - 6 - 7 577 71
BC051484.1 - 6 - 7 570 1380946 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001647.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Syf2

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
mRNA processing
mitotic G2 DNA damage checkpoint
gastrulation
RNA splicing
embryonic organ development
poly(A) RNA binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325610 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325607 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325609 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325606 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325608 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325611 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P535255 AY033432.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P535255 BC051484.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P535255 NM_026780.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P535255 AK003787.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P535255 AK152794.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P535255 AY033432.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P535255 BC051484.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P535255 NM_026780.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P535255 AK003787.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P535255 AK152794.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63751 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901736 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755217 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647526 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111780 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158309 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P535255 AY033432.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P535255 BC051484.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P535255 NM_026780.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P535255 AK003787.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P535255 AK152794.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P535255 AY033432.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P535255 BC051484.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P535255 NM_026780.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P535255 AK003787.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P535255 AK152794.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325591 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325598 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325594 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325588 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325600 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325607 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325611 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325602 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325616 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325608 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325604 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325596 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325605 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325614 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325613 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325606 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325590 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325593 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00995671 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05413204 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158309 - -
Whole Mouse Genome G4122A A_52_P535255 AY033432.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P535255 BC051484.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P535255 NM_026780.3 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P535255 AK003787.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P535255 AK152794.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P535255 AY033432.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P535255 BC051484.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P535255 NM_026780.3 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P535255 AK003787.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P535255 AK152794.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107376 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158309 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03995832 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03782196 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03995831 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03320733 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03320735 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03352648 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325608 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325606 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325609 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325611 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325607 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325610 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325649 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325679 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325561 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325641 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325606 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325596 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325617 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325564 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325622 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325628 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325581 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325563 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325660 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325656 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325566 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325673 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325569 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325558 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325591 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325618 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325604 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325550 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325629 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325608 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325634 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325579 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325616 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325555 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325571 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325678 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325652 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325576 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325574 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325620 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325567 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325630 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325625 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325680 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325582 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325670 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325556 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325688 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325675 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325657 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325599 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325624 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325611 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325575 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325613 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325676 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325648 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325682 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325638 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325687 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325621 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325665 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325645 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325685 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325636 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325593 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325560 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325667 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325647 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325588 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325577 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325551 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325678 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325648 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03667267 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03320733 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325620 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03753494 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325681 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03382577 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03702225 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03702557 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03896527 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325598 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03995831 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325669 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325628 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325688 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325602 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325599 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325590 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03753496 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325687 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325632 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03352649 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03388362 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325618 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325606 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325645 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325588 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325646 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03320738 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03320735 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03702229 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325638 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03995832 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03846727 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03702224 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325670 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325639 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03614245 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325596 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03614247 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03382578 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325675 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03753499 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325653 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325613 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325652 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325671 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325676 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03333610 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03614246 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325616 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03426843 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23325682 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03320736 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03388365 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03896528 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03382575 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10509028 - -
Affymetrix Mouse Gene 1.0 ST Array 10509025 - -
Affymetrix Mouse Gene 1.0 ST Array 10509027 - -
Affymetrix Mouse Gene 1.0 ST Array 10509024 - -
Affymetrix Mouse Gene 1.0 ST Array 10509026 - -
Affymetrix Mouse Gene 1.0 ST Array 10509029 - -
Affymetrix HT MG-430 PM Array Plate 1424104_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10509023 - -
Mouse Genome 430 2.0 Array 1424104_at - -
Mouse Genome 430 2.0 Array 1424104_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 68592_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 68592_PM_at - -
Affymetrix Mouse Clariom S Array PSR0400013147.mm.1 NM_026780.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400013147.mm.1 NM_026780.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400013147.mm.1 TC0400001647.mm.1 MTA TC
Mouse Expression Set 430 1424104_at - -
Mouse Expression Set 430 1424104_PM_at - -
Murine Genome U74v2 Set 96341_PM_at - -
Murine Genome U74v2 Set 96341_at - -
Mouse Genome 430A 2.0 Array 1424104_at - -
Mouse Genome 430A 2.0 Array 1424104_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1424104_at - -
Affymetrix HT Mouse Genome 430A Array 1424104_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 523258 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 360333 AJ292467.1 genbank_mrna_mouse
MouseWG-6 v2 Expression BeadChip 360333 AJ292468.1 genbank_mrna_mouse
MouseWG-6 v2 Expression BeadChip 360333 BC024797.1 genbank_mrna_mouse
MouseWG-6 v2 Expression BeadChip 360333 NM_001103179.1 refseq_mrna_mouse_NM
MouseWG-6 v2 Expression BeadChip 360333 NM_145125.3 refseq_mrna_mouse_NM
MouseWG-6 v2 Expression BeadChip 4070735 - -
Illumina mouse-6 v1.0 expression beadchip 360333 AJ292467.1 genbank_mrna_mouse
Illumina mouse-6 v1.0 expression beadchip 360333 AJ292468.1 genbank_mrna_mouse
Illumina mouse-6 v1.0 expression beadchip 360333 BC024797.1 genbank_mrna_mouse
Illumina mouse-6 v1.0 expression beadchip 360333 NM_001103179.1 refseq_mrna_mouse_NM
Illumina mouse-6 v1.0 expression beadchip 360333 NM_145125.3 refseq_mrna_mouse_NM
Illumina mouse-6 v1.0 expression beadchip 4070735 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004070735 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000360333 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1242551 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1216167 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1216167 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1242551 - -
mouse-6 v1.1 (Illumina) scl0002653.1_3-S - -
mouse-6 v1.1 (Illumina) scl24839.6.1_26-S - -
Illumina Sentrix mouse-6.v1 bead array 4070735 - -
Illumina Sentrix mouse-6.v1 bead array 360333 AJ292467.1 genbank_mrna_mouse
Illumina Sentrix mouse-6.v1 bead array 360333 AJ292468.1 genbank_mrna_mouse
Illumina Sentrix mouse-6.v1 bead array 360333 BC024797.1 genbank_mrna_mouse
Illumina Sentrix mouse-6.v1 bead array 360333 NM_001103179.1 refseq_mrna_mouse_NM
Illumina Sentrix mouse-6.v1 bead array 360333 NM_145125.3 refseq_mrna_mouse_NM
Illumina Sentrix MouseRef-8 Expression BeadChip scl24839.6.1_26-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0002653.1_3-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 360333 AJ292467.1 genbank_mrna_mouse
Illumina Sentrix Mouse-6 Expression BeadChip (2) 360333 AJ292468.1 genbank_mrna_mouse
Illumina Sentrix Mouse-6 Expression BeadChip (2) 360333 BC024797.1 genbank_mrna_mouse
Illumina Sentrix Mouse-6 Expression BeadChip (2) 360333 NM_001103179.1 refseq_mrna_mouse_NM
Illumina Sentrix Mouse-6 Expression BeadChip (2) 360333 NM_145125.3 refseq_mrna_mouse_NM
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4070735 - -
MouseRef-8 v2 Expression BeadChip ILMN_2956381 - -
MouseRef-8 v2 Expression BeadChip ILMN_1242551 - -
MouseRef-8 v2 Expression BeadChip ILMN_1216167 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1242551 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2956381 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1216167 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl24839.6.1_26-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0002653.1_3-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline