This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00503606_g1
See other TMEM25 GE Assays ›
Gene Symbol:
Tmem25
Gene Name:
transmembrane protein 25
Gene Aliases:
0610039J01Rik, AI429491
Chromosome Location:
Chr.9: 44793772 - 44799273 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00503606_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027865.2 NP_082141.1 4 - 5 732 111
GenBank mRNA AK002841.1 - 4 - 5 733 111
AK154206.1 - 3 - 4 1218 111
AK165199.1 - 4 - 5 764 111
AK165827.1 - 4 - 5 717 111
BC060057.1 - 4 - 5 752 5696102 111
BC060243.1 - 4 - 5 752 5696102 111

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002179.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmem25

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900018122.mm.1 TC0900002179.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900018124.mm.1 NM_027865.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900018124.mm.1 NM_027865.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900018124.mm.1 TC0900002179.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline