This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00503679_m1
See other PPP1R14D GE Assays ›
Gene Symbol:
Ppp1r14d
Gene Name:
protein phosphatase 1, regulatory (inhibitor) subunit 14D
Gene Aliases:
2010107K19Rik, GBPI-1, Ppp1r14
Chromosome Location:
Chr.2: 119218119 - 119230067 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00503679_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r14d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028104.4 NP_082380.1 1 - 2 325 94
GenBank mRNA AF408400.2 - 1 - 2 324 94
AK008348.1 - 1 - 2 283 94
BC024547.1 - 1 - 2 324 4989982 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004425.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp1r14d

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of protein dephosphorylation
regulation of phosphorylation
negative regulation of catalytic activity
protein phosphatase inhibitor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21568531 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21568534 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21568533 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009673 NM_028104.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009673 AK008348.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009673 BC024547.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 35473 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 820574 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 40579 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 851221 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 446763 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510094 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 546305 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 576452 AK039069.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 576452 AK039075.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 43250 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 173320 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121243 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139472 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009673 NM_028104.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009673 AK008348.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009673 BC024547.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568483 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568510 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568494 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568520 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568513 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568505 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568508 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568481 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568507 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568525 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568486 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568498 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568497 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568493 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568496 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568480 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568527 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568489 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568491 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568485 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568522 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568518 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568495 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568534 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568514 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568500 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568501 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568531 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568503 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568515 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568484 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568529 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568516 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139472 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121243 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04664598 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04664589 - -
Whole Mouse Genome G4122A A_52_P40345 NM_028104.3 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P40345 AF408400.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P40345 AK008348.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P40345 AY179331.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121243 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139472 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04664589 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01350840 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02259046 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01350825 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02259052 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02259047 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02259050 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01350824 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02259054 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01350842 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21568533 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21568534 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21568531 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568398 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568535 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568478 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568454 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568549 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568428 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568513 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568495 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568514 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568393 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568441 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568430 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568480 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568476 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568494 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568416 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568487 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568410 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568406 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568475 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568516 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568401 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568409 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568507 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568469 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568412 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568436 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568488 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568407 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568420 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568505 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568501 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568490 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568551 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568387 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568527 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568510 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568403 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568538 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568520 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568414 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568541 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568498 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568466 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568496 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568503 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568548 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568447 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568537 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568404 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568546 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568440 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568424 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568449 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568547 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568399 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568451 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568397 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568472 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568437 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568429 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568431 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568484 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568456 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568531 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568443 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568400 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568458 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568522 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568543 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568389 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568434 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568534 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568529 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568545 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568525 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568452 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568486 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568485 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568553 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568539 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568481 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568498 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01479279 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568471 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01970355 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568494 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568462 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568480 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568490 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568472 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568473 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568470 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01747716 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568507 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01151628 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568464 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568469 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568484 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568466 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568485 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568465 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01328088 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02259046 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568475 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568476 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568500 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02180012 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01566903 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568495 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568510 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21568511 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02259053 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01429611 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10486194 - -
Affymetrix Mouse Gene 1.0 ST Array 10486193 - -
Affymetrix Mouse Gene 1.0 ST Array 10486195 - -
Affymetrix Mouse Gene 1.0 ST Array 10486196 - -
Affymetrix Mouse Gene 1.0 ST Array 10486192 - -
Affymetrix HT MG-430 PM Array Plate 1417990_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10486191 - -
Mouse Genome 430 2.0 Array 1417990_at - -
Mouse Genome 430 2.0 Array 1417990_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 72112_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 72112_PM_at - -
Affymetrix Mouse Clariom S Array PSR0200036979.mm.1 NM_028104.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200036979.mm.1 TC0200004425.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200036981.mm.1 NM_028104.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200036981.mm.1 TC0200004425.mm.1 MTA TC
Mouse Expression Set 430 1417990_PM_at - -
Mouse Expression Set 430 1417990_at - -
Murine Genome U74v2 Set 165491_PM_at - -
Murine Genome U74v2 Set 165491_at - -
Mouse Genome 430A 2.0 Array 1417990_PM_at - -
Mouse Genome 430A 2.0 Array 1417990_at - -
Affymetrix HT Mouse Genome 430A Array 1417990_at - -
Affymetrix HT Mouse Genome 430A Array 1417990_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 911946 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 2190168 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002190168 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1253015 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1253015 - -
mouse-6 v1.1 (Illumina) scl18808.6.1_35-S - -
Illumina Sentrix mouse-6.v1 bead array 2190168 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl18808.6.1_35-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2190168 - -
MouseRef-8 v2 Expression BeadChip ILMN_2872371 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2872371 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1253015 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0