Assay Details
| Gene Symbol: Anln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028390.3 | NP_082666.2 | 16 - 17 | 2862 | 108 | |
| GenBank mRNA | AK045703.1 | - | 13 - 14 | 1826 | 108 | |
| AK046102.1 | - | 5 - 6 | 648 | 108 | ||
| BC030489.1 | - | 9 - 10 | 1204 | 5373071 | 108 | |
| BC032164.1 | - | 16 - 17 | 2880 | 5346703 | 108 | |
More Information
Set Membership: |
|

Anln
Genomic Map