This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00504689_g1
See other RBM39 GE Assays ›
Gene Symbol:
Rbm39
Gene Name:
RNA binding motif protein 39
Gene Aliases:
1500012C14Rik, 2310040E03Rik, B330012G18Rik, C79248, R75070, Rnpc2, caper
Chromosome Location:
Chr.2: 156147239 - 156180238 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00504689_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbm39
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291114.1 NP_001278043.1 4 - 5 662 58
NM_001291115.1 NP_001278044.1 4 - 5 457 58
NM_133242.3 NP_573505.2 4 - 5 662 58
GenBank mRNA AK135289.1 - 3 - 4 288 58
AK146046.1 - 4 - 5 457 58
AK147076.1 - 4 - 5 607 58
AK154989.1 - 4 - 5 593 58
BC023205.1 - 4 - 5 564 58
BC030493.1 - 2 - 3 277 5400675 58
BC082607.1 - 2 - 3 252 30667785 58
BC086645.1 - 4 - 5 585 6817169 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004983.mm.1

Additional Information:

This assay is in the 5'UTR region of the BC082607.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbm39

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
RNA splicing
regulation of nucleic acid-templated transcription
nucleotide binding
nucleic acid binding
transcription coactivator activity
RNA binding
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200042589.mm.1 TC0200004983.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200042573.mm.1 NM_133242.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200042573.mm.1 TC0200004983.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200042585.mm.1 NM_133242.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200042585.mm.1 TC0200004983.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0