This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00505396_m1
See other STARD4 GE Assays ›
Gene Symbol:
Stard4
Gene Name:
StAR-related lipid transfer (START) domain containing 4
Gene Aliases:
4632419C16Rik, 9030213J02Rik, AA517649, AW324468
Chromosome Location:
Chr.18: 33199140 - 33213879 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00505396_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stard4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133774.4 NP_598535.1 3 - 4 346 100
GenBank mRNA AF480297.1 - 3 - 4 201 100
AF480298.1 - 3 - 4 201 100
AK042347.1 - 3 - 4 350 100
AK042691.1 - 3 - 4 306 100
AK054501.1 - 3 - 4 347 100
AK078858.1 - 3 - 4 350 100
AK085331.1 - 3 - 4 350 100
AK165641.1 - 3 - 4 352 100
BC005642.1 - 3 - 4 346 3709232 100

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stard4

Gene Ontology Categories:

Function(s) Process(es)

transport
lipid transport
lipid binding
cholesterol binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline