This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00505929_g1
See other UNC45A GE Assays ›
Gene Symbol:
Unc45a
Gene Name:
unc-45 homolog A (C. elegans)
Gene Aliases:
AW538196
Chromosome Location:
Chr.7: 80325292 - 80341000 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00505929_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Unc45a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133952.2 NP_598713.2 3 - 4 345 76
GenBank mRNA AK047292.1 - 3 - 4 292 76
AK047476.1 - 3 - 4 292 76
AK088034.1 - 3 - 4 297 76
AK166809.1 - 3 - 4 328 76
AK202434.1 - 2 - 3 120 76
BC004717.1 - 3 - 4 334 3582116 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003591.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Unc45a

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
muscle organ development
cell differentiation
chaperone-mediated protein folding
protein binding
Hsp90 protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700030381.mm.1 NM_133952.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700030381.mm.1 NM_133952.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700030381.mm.1 TC0700003591.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline