This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00505939_g1
See other UNC45A GE Assays ›
Gene Symbol:
Unc45a
Gene Name:
unc-45 homolog A (C. elegans)
Gene Aliases:
AW538196
Chromosome Location:
Chr.7: 80325292 - 80341000 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00505939_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Unc45a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133952.2 NP_598713.2 14 - 15 2104 66
GenBank mRNA AK028912.1 - 14 - 15 2090 66
AK047292.1 - 14 - 15 2051 66
AK047476.1 - 14 - 15 2051 66
AK088034.1 - 14 - 15 2025 66
AK166809.1 - 14 - 15 2087 66
BC004717.1 - 14 - 15 2093 3582116 66

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Unc45a

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
muscle organ development
cell differentiation
chaperone-mediated protein folding
protein binding
Hsp90 protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0