This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00506765_g1
See other APOBR GE Assays ›
Gene Symbol:
Apobr
Gene Name:
apolipoprotein B receptor
Gene Aliases:
Apob-48r, Apob48r
Chromosome Location:
Chr.7: 126584912 - 126589125 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00506765_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apobr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138310.1 NP_612183.1 1 - 2 64 77
GenBank mRNA AF141335.1 - 1 - 2 64 77
AK156422.1 - 1 - 2 131 77
BC030718.1 - 1 - 2 72 4482457 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Apobr

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
triglyceride metabolic process
transport
lipid transport
receptor-mediated endocytosis
steroid metabolic process
cholesterol metabolic process
lipoprotein particle receptor activity
very-low-density lipoprotein particle receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline