This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00507242_g1
See other ABCG4 GE Assays ›
Gene Symbol:
Abcg4
Gene Name:
ATP-binding cassette, sub-family G (WHITE), member 4
Gene Aliases:
6430517O04Rik
Chromosome Location:
Chr.9: 44273188 - 44289102 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00507242_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Abcg4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138955.3 NP_620405.3 1 - 2 233 76
GenBank mRNA AF411084.1 - 1 - 2 111 76
AF425077.1 - 1 - 2 113 76
AJ426047.2 - 1 - 2 112 76
AK043147.1 - 1 - 2 234 76
AK157317.1 - 1 - 2 558 76
AY040865.1 - 1 - 2 77 76
BC016200.1 - 1 - 2 113 4511456 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002153.mm.1

Additional Information:

This assay is in the 5'UTR region of the AF411084.1,AF425077.1,AJ426047.2,AK043147.1,AK157317.1,AY040865.1,BC016200.1,NM_138955.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Abcg4

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

cholesterol efflux
protein homodimerization activity
protein heterodimerization activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900017883.mm.1 NM_138955.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900017883.mm.1 NM_138955.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900017883.mm.1 TC0900002153.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900017876.mm.1 NM_138955.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900017876.mm.1 NM_138955.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900017876.mm.1 TC0900002153.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900017881.mm.1 NM_138955.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900017881.mm.1 NM_138955.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900017881.mm.1 TC0900002153.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 459022 - -
ABI Mouse Genome Survey Microarray 929858 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0