This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00507251_g1
See other ABCG4 GE Assays ›
Gene Symbol:
Abcg4
Gene Name:
ATP-binding cassette, sub-family G (WHITE), member 4
Gene Aliases:
6430517O04Rik
Chromosome Location:
Chr.9: 44273188 - 44289102 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00507251_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Abcg4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138955.3 NP_620405.3 10 - 11 1417 95
GenBank mRNA AF378330.1 - 9 - 10 1248 95
AF411084.1 - 10 - 11 1295 95
AF425077.1 - 10 - 11 1297 95
AJ426047.2 - 10 - 11 1296 95
AK043147.1 - 10 - 11 1418 95
AK138415.1 - 9 - 10 1258 95
AK157317.1 - 10 - 11 1742 95
AY040865.1 - 10 - 11 1261 95
BC016200.1 - 10 - 11 1297 4511456 95
BC023077.1 - 4 - 5 505 95
BC026477.1 - 632 95
BC059033.1 - 1 - 2 590 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002153.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Abcg4

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

cholesterol efflux
protein homodimerization activity
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900017858.mm.1 NM_138955.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900017858.mm.1 NM_138955.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900017858.mm.1 TC0900002153.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0