Assay Details
| Gene Symbol: Aox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023631.2 | NP_076120.2 | 3 - 4 | 388 | 75 | |
| GenBank mRNA | AF233581.1 | - | 3 - 4 | 315 | 75 | |
| AK158427.1 | - | 3 - 4 | 389 | 75 | ||
| AK160953.1 | - | 3 - 4 | 354 | 75 | ||
| BC117975.1 | - | 3 - 4 | 315 | 40098789 | 75 | |
| BC117976.1 | - | 3 - 4 | 315 | 40098791 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aox4
Genomic Map