This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00508241_mH
See other ASF1B GE Assays ›
Gene Symbol:
Asf1b
Gene Name:
anti-silencing function 1B histone chaperone
Gene Aliases:
1700003K02Rik, AA409591
Chromosome Location:
Chr.8: 83955694 - 83970195 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00508241_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Asf1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024184.2 NP_077146.1 1 - 2 245 71
GenBank mRNA AK005646.1 - 1 - 2 278 71
AK077005.1 - 1 - 2 271 71
AK077718.1 - 1 - 2 226 71
AK134906.1 - 1 - 2 433 71
AK150818.1 - 1 - 2 226 71
AK151951.1 - 1 - 2 226 71
AK165322.1 - 1 - 2 224 71
BC003428.1 - 1 - 2 245 2645378 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000937.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Asf1b

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

chromatin assembly or disassembly
nucleosome assembly
DNA replication-dependent nucleosome assembly
DNA replication-independent nucleosome assembly
transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
histone binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800006967.mm.1 NM_024184.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006967.mm.1 NM_024184.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800006967.mm.1 TC0800000937.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800006964.mm.1 NM_024184.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006964.mm.1 NM_024184.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800006964.mm.1 TC0800000937.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800006962.mm.1 NM_024184.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006962.mm.1 NM_024184.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800006962.mm.1 TC0800000937.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0