This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00508283_g1
See other DPY30 GE Assays ›
Gene Symbol:
Dpy30
Gene Name:
dpy-30 homolog (C. elegans)
Gene Aliases:
2810410M20Rik, C87842
Chromosome Location:
Chr.17: 74299474 - 74323944 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00508283_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dpy30
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001146223.1 NP_001139695.1 1 - 2 91 68
GenBank mRNA AK013073.1 - 1 - 2 80 68
AK077902.1 - 1 - 2 79 68
BC002240.1 - 1 - 2 60 3491912 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700002534.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK013073.1,AK077902.1,BC002240.1,NM_001146223.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
endosomal transport
chromatin modification
histone H3-K4 methylation
identical protein binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700023217.mm.1 NM_001146222.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700023217.mm.1 NM_001146222.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700023217.mm.1 NM_001146223.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700023217.mm.1 NM_001146223.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700023217.mm.1 TC1700002534.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700023219.mm.1 NM_001146223.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700023219.mm.1 NM_001146223.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700023219.mm.1 TC1700002534.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline