This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00508502_m1
See other PSMG3 GE Assays ›
Gene Symbol:
Psmg3
Gene Name:
proteasome (prosome, macropain) assembly chaperone 3
Gene Aliases:
1810042K04Rik, 4930403H09Rik, AI303239
Chromosome Location:
Chr.5: 139823594 - 139827003 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00508502_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Psmg3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025604.3 NP_079880.1 2 - 3 361 83
GenBank mRNA AK012616.1 - 2 - 3 362 83
BC027410.1 - 2 - 3 274 4237608 83
BC058692.1 - 2 - 3 347 6818127 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003743.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Psmg3

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24137874 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24137873 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24137875 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P289987 NM_025604.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P289987 AK012616.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P289987 BC027410.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P289987 BC058692.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P289987 NM_025604.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P289987 AK012616.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P289987 BC027410.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P289987 BC058692.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13724 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 938909 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174943 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P289987 NM_025604.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P289987 AK012616.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P289987 BC027410.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P289987 BC058692.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P289987 NM_025604.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P289987 AK012616.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P289987 BC027410.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P289987 BC058692.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24137887 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24137876 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24137867 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24137893 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24137886 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24137879 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24137878 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24137865 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24137892 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24137882 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24137890 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24137875 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24137891 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24137869 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24137874 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24137881 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24137872 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174943 - -
Whole Mouse Genome G4122A A_51_P289987 NM_025604.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P289987 AK012616.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P289987 BC027410.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P289987 BC058692.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P289987 NM_025604.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P289987 AK012616.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P289987 BC027410.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P289987 BC058692.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131900 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174943 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04740050 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04740049 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24137874 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24137875 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24137873 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137891 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137792 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137809 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137872 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137887 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137788 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137890 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137826 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137814 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137827 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137859 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137803 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137865 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137903 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137879 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137869 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137892 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137881 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137824 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137896 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137797 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137799 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137861 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137883 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137815 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137791 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137810 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137893 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137822 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137905 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137867 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137886 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137875 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137796 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137805 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137899 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137813 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137820 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137817 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24137874 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137881 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137872 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137891 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04360498 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137863 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137874 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04587569 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137895 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137865 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137876 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137878 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04523002 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137875 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04482428 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04445565 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137893 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137882 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137877 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137859 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04573414 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137890 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137867 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04522995 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137899 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137879 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04882019 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137887 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04594398 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24137886 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10535185 - -
Affymetrix Mouse Gene 1.0 ST Array 10535187 - -
Affymetrix Mouse Gene 1.0 ST Array 10535186 - -
Affymetrix HT MG-430 PM Array Plate 1424087_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1430572_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10535184 - -
Mouse Genome 430 2.0 Array 1430572_at - -
Mouse Genome 430 2.0 Array 1424087_PM_at - -
Mouse Genome 430 2.0 Array 1430572_PM_at - -
Mouse Genome 430 2.0 Array 1424087_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66506_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66506_PM_at - -
Affymetrix Mouse Clariom S Array PSR0500032098.mm.1 TC0500003743.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500032098.mm.1 NM_025604.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500032098.mm.1 NM_025604.3 Refseq
Mouse Expression Set 430 1424087_PM_at - -
Mouse Expression Set 430 1424087_at - -
Mouse Expression Set 430 1430572_at - -
Mouse Expression Set 430 1430572_PM_at - -
Murine Genome U74v2 Set 162356_PM_r_at - -
Murine Genome U74v2 Set 94543_at - -
Murine Genome U74v2 Set 94543_PM_at - -
Murine Genome U74v2 Set 162356_r_at - -
Murine Genome U74v2 Set 167279_PM_at - -
Murine Genome U74v2 Set 167279_at - -
Mouse Genome 430A 2.0 Array 1424087_PM_at - -
Mouse Genome 430A 2.0 Array 1424087_at - -
Affymetrix HT Mouse Genome 430A Array 1424087_at - -
Affymetrix HT Mouse Genome 430A Array 1424087_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1430572_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1430572_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 332838 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 6350358 - -
Illumina mouse-6 v1.0 expression beadchip 6350358 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006350358 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2695599 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2695599 - -
mouse-6 v1.1 (Illumina) scl066506.1_15-S - -
Illumina Sentrix mouse-6.v1 bead array 6350358 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl066506.1_15-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6350358 - -
MouseRef-8 v2 Expression BeadChip ILMN_2695599 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2695599 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0