This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00508528_g1
See other OGFOD2 GE Assays ›
Gene Symbol:
Ogfod2
Gene Name:
2-oxoglutarate and iron-dependent oxygenase domain containing 2
Gene Aliases:
1300006G11Rik, 5730405M13Rik, AU041636, AW552395
Chromosome Location:
Chr.5: 124112058 - 124115480 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00508528_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ogfod2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025671.2 NP_079947.1 5 - 6 525 61
GenBank mRNA AK004943.1 - 5 - 6 526 61
AK017497.1 - 5 - 6 526 61
AK018534.1 - 5 - 6 526 61
BC022762.1 - 4 - 5 373 61
BC069872.1 - 5 - 6 528 4219293 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001436.mm.1

Additional Information:

This assay is in the 5'UTR region of the BC022762.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ogfod2

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
sulfonate dioxygenase activity
molecular_function
iron ion binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
2,4-dichlorophenoxyacetate alpha-ketoglutarate dioxygenase activity
procollagen-proline dioxygenase activity
L-ascorbic acid binding
hypophosphite dioxygenase activity
DNA-N1-methyladenine dioxygenase activity
metal ion binding
dioxygenase activity
C-19 gibberellin 2-beta-dioxygenase activity
C-20 gibberellin 2-beta-dioxygenase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500012868.mm.1 TC0500001436.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500012868.mm.1 NM_025671.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500012868.mm.1 NM_025671.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500012869.mm.1 TC0500001436.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500012869.mm.1 NM_025671.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500012869.mm.1 NM_025671.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-tlmfb:80/100.66.76.102:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0