This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00509018_m1
See other DRAM2 GE Assays ›
Gene Symbol:
Dram2
Gene Name:
DNA-damage regulated autophagy modulator 2
Gene Aliases:
2010305N14Rik, 2610318G18Rik, AI647667, Tmem77
Chromosome Location:
Chr.3: 106547798 - 106575341 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00509018_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dram2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025582.2 NP_001020753.1 2 - 3 250 99
NM_026013.3 NP_080289.1 2 - 3 250 99
GenBank mRNA AI875295.1 - 2 - 3 250 99
AK008532.1 - 2 - 3 174 99
AK009940.1 - 2 - 3 187 99
AK012044.1 - 2 - 3 167 97
AK172455.1 - 2 - 3 217 99
BC024458.1 - 2 - 3 176 4975302 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001104.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK008532.1,AK009940.1,AK012044.1,AK172455.1,BC024458.1,NM_001025582.2,NM_026013.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dram2

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
regulation of autophagy
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300008986.mm.1 NM_001025582.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300008986.mm.1 NM_001286986.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008986.mm.1 NM_001286987.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008986.mm.1 NM_026013.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300008986.mm.1 TC0300001104.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300008973.mm.1 NM_001025582.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300008973.mm.1 NM_001286986.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008973.mm.1 NM_001286987.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008973.mm.1 NM_026013.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300008973.mm.1 TC0300001104.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300008981.mm.1 NM_001025582.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300008981.mm.1 NM_001286986.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008981.mm.1 NM_001286987.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008981.mm.1 NM_026013.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300008981.mm.1 TC0300001104.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300008984.mm.1 NM_001025582.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300008984.mm.1 NM_001286986.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008984.mm.1 NM_026013.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300008984.mm.1 TC0300001104.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 906617 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w64qk:80/100.66.78.26:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0