This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00509468_g1
See other IAH1 GE Assays ›
Gene Symbol:
Iah1
Gene Name:
isoamyl acetate-hydrolyzing esterase 1 homolog (S. cerevisiae)
Gene Aliases:
1500019E20Rik, 4833421E05Rik
Chromosome Location:
Chr.12: 21316360 - 21323607 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00509468_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Iah1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026347.3 NP_080623.2 2 - 3 167 116
GenBank mRNA AK005287.1 - 2 - 3 168 116
AK011299.1 - 2 - 3 168 116
AK014743.1 - 2 - 3 145 116
BC060949.1 - 2 - 3 170 6533986 116
BC087901.1 - 2 - 3 138 30303437 116

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200000183.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Iah1

Panther Classification:

Molecular Function -

esterase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
biological_process
lipid catabolic process
molecular_function
nuclease activity
hydrolase activity
hydrolase activity, acting on ester bonds
phosphoric ester hydrolase activity
T/G mismatch-specific endonuclease activity
retroviral 3' processing activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200001467.mm.1 NM_026347.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200001467.mm.1 NM_026347.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200001467.mm.1 TC1200000183.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0