This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00509621_m1
See other BPHL GE Assays ›
Gene Symbol:
Bphl
Gene Name:
biphenyl hydrolase-like (serine hydrolase, breast epithelial mucin-associated antigen)
Gene Aliases:
2010012D11Rik, 5730533B08Rik, AI115341
Chromosome Location:
Chr.13: 34037593 - 34074074 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00509621_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bphl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026512.1 NP_080788.1 2 - 3 234 63
GenBank mRNA AK002907.1 - 2 - 3 232 63
AK008202.1 - 2 - 3 63
AK017801.1 - 2 - 3 237 63
AK078232.1 - 2 - 3 279 63
AK152712.1 - 2 - 3 244 63
AK170099.1 - 2 - 3 237 63
BC025162.1 - 2 - 3 234 4921451 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000385.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bphl

Panther Classification:

Molecular Function -

hydrolase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
nuclease activity
hydrolase activity
phosphoric ester hydrolase activity
T/G mismatch-specific endonuclease activity
retroviral 3' processing activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300002335.mm.1 NM_026512.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300002335.mm.1 NM_026512.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300002335.mm.1 TC1300000385.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline