This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00509906_g1
See other HAGHL GE Assays ›
Gene Symbol:
Haghl
Gene Name:
hydroxyacylglutathione hydrolase-like
Gene Aliases:
1500017E18Rik, 2810014I23Rik, C330022E15Rik
Chromosome Location:
Chr.17: 25779843 - 25785669 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00509906_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Haghl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271433.1 NP_001258362.1 7 - 8 848 71
NM_001271435.1 NP_001258364.1 7 - 8 848 71
NM_001271436.1 NP_001258365.1 6 - 7 662 71
NM_001271437.1 NP_001258366.1 6 - 7 657 71
NM_026897.3 NP_081173.1 6 - 7 833 71
GenBank mRNA AK005274.1 - 6 - 7 829 71
AK021220.1 - 6 - 7 664 71
AK047517.1 - 7 - 8 803 71
AK079632.1 - 4 - 5 997 71
AK154248.1 - 6 - 7 638 71
BC030466.1 - 7 - 8 833 5253213 71
BC083322.1 - 7 - 8 787 6820885 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001672.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Haghl

Gene Ontology Categories:

Function(s) Process(es)

glutathione biosynthetic process
biological_process
molecular_function
hydroxyacylglutathione hydrolase activity
zinc ion binding
CoA hydrolase activity
hydrolase activity
2-oxoglutaryl-CoA thioesterase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA thioesterase activity
3-isopropylbut-3-enoyl-CoA thioesterase activity
glutaryl-CoA hydrolase activity
metal ion binding
acyl-CoA hydrolase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700015273.mm.1 NM_001271433.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015273.mm.1 NM_001271434.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015273.mm.1 NM_001271435.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015273.mm.1 NM_001271436.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015273.mm.1 NM_001271437.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015273.mm.1 NM_026897.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700015273.mm.1 TC1700001672.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700015260.mm.1 NM_001271435.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015260.mm.1 TC1700001672.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700015274.mm.1 NM_001271433.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015274.mm.1 NM_001271434.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015274.mm.1 NM_001271435.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015274.mm.1 NM_001271436.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015274.mm.1 NM_001271437.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015274.mm.1 NM_026897.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700015274.mm.1 TC1700001672.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700015278.mm.1 NM_001271433.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015278.mm.1 NM_001271434.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015278.mm.1 NM_001271435.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015278.mm.1 NM_001271436.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015278.mm.1 NM_001271437.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015278.mm.1 NM_026897.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700015278.mm.1 TC1700001672.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 367267 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline