This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00510341_m1
See other PPIL3 GE Assays ›
Gene Symbol:
Ppil3
Gene Name:
peptidylprolyl isomerase (cyclophilin)-like 3
Gene Aliases:
2310076N22Rik, 2510026K04Rik, Cyp10l
Chromosome Location:
Chr.1: 58430993 - 58445527 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00510341_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppil3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285826.1 NP_001272755.1 3 - 4 802 118
NM_001285827.1 NP_001272756.1 3 - 4 467 118
NM_027351.3 NP_081627.1 3 - 4 281 118
NM_027374.3 NP_081650.2 3 - 4 467 118
GenBank mRNA AK010207.1 - 3 - 4 285 118
AK011000.1 - 3 - 4 236 118
BC060993.1 - 3 - 4 249 6591278 118
BC061645.1 - 3 - 4 745 5032617 118
CJ166559.1 - 3 - 4 411 118
DV043111.1 - 3 - 4 503 118

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002382.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppil3

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
mRNA processing
protein folding
RNA splicing
peptidyl-prolyl cis-trans isomerase activity
isomerase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100019378.mm.1 NM_001285826.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100019378.mm.1 NM_001285827.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100019378.mm.1 NM_027351.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100019378.mm.1 NM_027351.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100019378.mm.1 NM_027374.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100019378.mm.1 NM_027374.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100019378.mm.1 TC0100002382.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline