This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00510380_m1
See other POLR3F GE Assays ›
Gene Symbol:
Polr3f
Gene Name:
polymerase (RNA) III (DNA directed) polypeptide F
Gene Aliases:
2810411G20Rik, 3010019O03Rik, 3110032A07Rik, RPC39, RPC6
Chromosome Location:
Chr.2: 144527745 - 144541779 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00510380_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr3f
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029763.3 NP_084039.2 1 - 2 172 92
GenBank mRNA AK013084.1 - 1 - 2 144 92
AK013897.1 - 1 - 2 159 92
AK014110.1 - 1 - 2 159 92
AK029286.1 - 1 - 2 173 92
AK036166.1 - 1 - 2 169 92
AK076538.1 - 1 - 2 199 92
AK144679.1 - 1 - 2 170 92
BC009159.1 - 1 - 2 161 3602180 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002149.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK013084.1,AK029286.1,AK036166.1,AK076538.1,AK144679.1,BC009159.1,NM_029763.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polr3f

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

immune system process
transcription, DNA-templated
positive regulation of interferon-beta production
innate immune response
positive regulation of innate immune response
defense response to virus
RNA polymerase III activity
DNA binding
DNA-directed RNA polymerase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200016941.mm.1 NM_029763.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200016941.mm.1 NM_029763.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200016941.mm.1 TC0200002149.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200016943.mm.1 NM_029763.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200016943.mm.1 NM_029763.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200016943.mm.1 TC0200002149.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 453393 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0