This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00510837_g1
See other GTF3C2 GE Assays ›
Gene Symbol:
Gtf3c2
Gene Name:
general transcription factor IIIC, polypeptide 2, beta
Gene Aliases:
1300004C11Rik, 2610510G03Rik, AI225816, AU041069, TFIIIC110, mKIAA0011
Chromosome Location:
Chr.5: 31156005 - 31180658 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00510837_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf3c2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027901.2 NP_082177.2 14 - 15 2311 77
GenBank mRNA AK004895.1 - 14 - 15 2089 77
AK012095.1 - 7 - 8 908 77
AK083585.1 - 14 - 15 2313 77
AK083636.1 - 13 - 14 2066 77
AK129033.1 - 13 - 14 2061 77
AK138760.1 - 2080 77
AK149771.1 - 15 - 16 2208 77
AK151300.1 - 14 - 15 2076 77
AK152439.1 - 14 - 15 2076 77
AK159938.1 - 14 - 15 2079 77
BC026573.1 - 6 - 7 629 77
BC034369.1 - 14 - 15 2089 3994677 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003724.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf3c2

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
transcription from RNA polymerase III promoter
DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500019840.mm.1 TC0500003724.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500019840.mm.1 NM_027901.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500019840.mm.1 NM_027901.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500019839.mm.1 TC0500003724.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500019839.mm.1 NM_027901.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500019839.mm.1 NM_027901.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0