This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00511022_g1
See other CPSF3L GE Assays ›
Gene Symbol:
Cpsf3l
Gene Name:
cleavage and polyadenylation specific factor 3-like
Gene Aliases:
2410006F12Rik, AU044035
Chromosome Location:
Chr.4: 155869548 - 155889103 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00511022_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cpsf3l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028020.3 NP_082296.1 12 - 13 1426 63
GenBank mRNA AK010425.1 - 12 - 13 1388 63
AK090206.1 - 12 - 13 1427 63
AK150436.1 - 12 - 13 1423 63
AK152740.1 - 12 - 13 1423 63
AK167607.1 - 12 - 13 1429 63
AK172533.1 - 12 - 13 1410 63
BC008240.1 - 4 - 5 522 3601302 63
BC011155.1 - 12 - 13 1410 4195656 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002097.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cpsf3l

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

snRNA processing
molecular_function
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400017571.mm.1 NM_028020.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017571.mm.1 NM_028020.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400017571.mm.1 TC0400002097.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400017570.mm.1 NM_028020.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017570.mm.1 NM_028020.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400017570.mm.1 TC0400002097.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline