This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00511140_m1
See other TRMU GE Assays ›
Gene Symbol:
Trmu
Gene Name:
tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase
Gene Aliases:
1110005N20Rik, 1600025P05Rik, AI314320, Trmt1
Chromosome Location:
Chr.15: 85879309 - 85897394 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00511140_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trmu
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028063.2 NP_082339.1 1 - 2 136 69
GenBank mRNA AK005541.1 - 1 - 2 136 69
AK043532.1 - 1 - 2 143 69
AK044059.1 - 1 - 2 143 69
AY349617.1 - 1 - 2 143 69
BC051548.1 - 1 - 2 139 69
BC061026.1 - 1 - 2 123 30305575 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000827.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trmu

Panther Classification:

Molecular Function -

RNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

tRNA wobble position uridine thiolation
tRNA processing
tRNA methylation
tRNA binding
nucleotide binding
molecular_function
RNA binding
tRNA (5-methylaminomethyl-2-thiouridylate)-methyltransferase activity
ATP binding
transferase activity
sulfurtransferase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500006505.mm.1 NM_028063.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500006505.mm.1 NM_028063.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500006505.mm.1 TC1500000827.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500006509.mm.1 NM_028063.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500006509.mm.1 NM_028063.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500006509.mm.1 TC1500000827.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500006506.mm.1 NM_028063.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500006506.mm.1 NM_028063.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500006506.mm.1 TC1500000827.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 930035 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0