This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00511146_m1
See other TRMU GE Assays ›
Gene Symbol:
Trmu
Gene Name:
tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase
Gene Aliases:
1110005N20Rik, 1600025P05Rik, AI314320, Trmt1
Chromosome Location:
Chr.15: 85879309 - 85897394 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00511146_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trmu
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028063.2 NP_082339.1 7 - 8 822 88
GenBank mRNA AK005541.1 - 7 - 8 822 88
AK043532.1 - 7 - 8 829 88
AK044059.1 - 7 - 8 829 88
AY349617.1 - 7 - 8 829 88
BC051548.1 - 7 - 8 823 88
BC061026.1 - 7 - 8 809 30305575 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000827.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trmu

Panther Classification:

Molecular Function -

RNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

tRNA wobble position uridine thiolation
tRNA processing
tRNA methylation
tRNA binding
nucleotide binding
molecular_function
RNA binding
tRNA (5-methylaminomethyl-2-thiouridylate)-methyltransferase activity
ATP binding
transferase activity
sulfurtransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500006528.mm.1 NM_028063.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500006528.mm.1 NM_028063.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500006528.mm.1 TC1500000827.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0