This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00511148_g1
See other TRMU GE Assays ›
Gene Symbol:
Trmu
Gene Name:
tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase
Gene Aliases:
1110005N20Rik, 1600025P05Rik, AI314320, Trmt1
Chromosome Location:
Chr.15: 85879309 - 85897394 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00511148_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trmu
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028063.2 NP_082339.1 9 - 10 1074 60
GenBank mRNA AK005541.1 - 9 - 10 1074 60
AK043532.1 - 9 - 10 1081 60
AK044059.1 - 9 - 10 1081 60
AY349617.1 - 9 - 10 1081 60
BC037371.1 - 3 - 4 329 60
BC048925.1 - 1 - 2 1630 60
BC051548.1 - 9 - 10 1075 60
BC061026.1 - 9 - 10 1061 30305575 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000827.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trmu

Panther Classification:

Molecular Function -

RNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

tRNA wobble position uridine thiolation
tRNA processing
tRNA methylation
tRNA binding
nucleotide binding
molecular_function
RNA binding
tRNA (5-methylaminomethyl-2-thiouridylate)-methyltransferase activity
ATP binding
transferase activity
sulfurtransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500006533.mm.1 NM_028063.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500006533.mm.1 NM_028063.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500006533.mm.1 TC1500000827.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0