This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00511229_m1
See other CHAF1B GE Assays ›
Gene Symbol:
Chaf1b
Gene Name:
chromatin assembly factor 1, subunit B (p60)
Gene Aliases:
2600017H24Rik, C76145, CAF-I p60, CAF-Ip60, CAF1, CAF1A, CAF1P60, MPHOSPH7
Chromosome Location:
Chr.16: 93883895 - 93906115 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00511229_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Chaf1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028083.4 NP_082359.1 11 - 12 1186 97
GenBank mRNA AK011243.1 - 11 - 12 1187 97
BC013532.1 - 10 - 11 1096 4238801 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001050.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Chaf1b

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
DNA replication
DNA repair
nucleosome assembly
DNA replication-dependent nucleosome assembly
transcription, DNA-templated
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
cell cycle
chromatin assembly
mitophagy in response to mitochondrial depolarization
protein binding
histone binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600008199.mm.1 NM_028083.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600008199.mm.1 NM_028083.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600008199.mm.1 TC1600001050.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600008198.mm.1 NM_028083.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600008198.mm.1 NM_028083.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600008198.mm.1 TC1600001050.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-s8qkh:80/100.66.76.99:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0