This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00511450_m1
See other TMEM175 GE Assays ›
Gene Symbol:
Tmem175
Gene Name:
transmembrane protein 175
Gene Aliases:
3010001K23Rik, AI504381
Chromosome Location:
Chr.5: 108629801 - 108647770 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00511450_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem175
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163532.1 NP_001157004.1 3 - 4 380 99
NM_028223.3 NP_082499.3 3 - 4 487 99
GenBank mRNA AI526594.1 - 3 - 4 351 99
AK013879.1 - 3 - 4 310 99
AK038865.1 - 3 - 4 347 99
AK049952.1 - 3 - 4 421 99
AK082765.1 - 3 - 4 343 99
AK170273.1 - 3 - 4 330 99
BB653267.1 - 3 - 4 417 99
BC005542.1 - 3 - 4 295 3590497 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001110.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmem175

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500009463.mm.1 TC0500001110.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500009463.mm.1 NM_001163531.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009463.mm.1 NM_001163531.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009463.mm.1 NM_001163532.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009463.mm.1 NM_001163532.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009463.mm.1 NM_028223.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009463.mm.1 NM_028223.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0