This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00511457_m1
See other TMEM175 GE Assays ›
Gene Symbol:
Tmem175
Gene Name:
transmembrane protein 175
Gene Aliases:
3010001K23Rik, AI504381
Chromosome Location:
Chr.5: 108629801 - 108647770 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00511457_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem175
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163531.1 NP_001157003.1 10 - 11 1032 75
NM_001163532.1 NP_001157004.1 10 - 11 1035 75
NM_028223.3 NP_082499.3 10 - 11 1142 75
GenBank mRNA AK013879.1 - 10 - 11 965 75
AK034579.1 - 10 - 11 1011 75
AK082765.1 - 10 - 11 1003 75
AK170273.1 - 10 - 11 985 75
BC005542.1 - 10 - 11 950 3590497 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001110.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmem175

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23973162 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2416862 AK076446.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989573 BC005542.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989573 AK082765.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989573 AK170273.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989573 AK034579.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989573 NM_028223.3 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 595833 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 623859 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 88251 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539957 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 966394 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 316482 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 641710 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 700197 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 657638 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 652428 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 695260 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 19019 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 94585 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 927 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119069 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135062 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989573 BC005542.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989573 AK082765.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989573 AK170273.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989573 AK034579.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989573 NM_028223.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2416862 AK076446.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23973168 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23973163 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23973166 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23973172 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23973167 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23973176 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23973171 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23973174 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23973169 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23973165 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23973164 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23973170 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23973177 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119069 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05690247 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135062 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05690258 - -
Whole Mouse Genome G4122A A_52_P548000 AK038865.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P548000 AK049952.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P456098 AK132151.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P456098 BC005542.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P456098 AK034579.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P456098 NM_028223.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P456098 NM_001163531.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132821 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119069 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135062 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04600428 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04600427 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04600454 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23973162 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973290 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973201 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973161 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973330 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973193 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973279 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973282 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973210 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973259 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973170 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973315 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973345 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973371 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973293 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973325 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973276 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973313 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973251 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973367 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973223 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973233 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973220 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973306 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973340 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973329 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973190 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973321 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973299 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973327 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973342 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973222 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973287 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973318 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973354 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973225 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973240 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973359 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973248 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973271 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973369 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973237 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973297 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973303 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973339 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973167 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973252 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973262 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973273 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973177 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973235 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973169 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973230 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973292 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973322 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973357 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973316 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973310 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973163 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973188 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973284 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973360 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973179 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973346 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973217 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973165 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973352 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973305 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973231 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973255 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973168 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973218 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973246 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973347 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973309 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973257 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973350 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973312 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973229 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973265 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973365 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973185 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973195 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973249 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973362 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973212 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973227 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973268 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973186 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973332 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973277 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973214 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973199 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23973335 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23973173 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23973170 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23973161 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04882131 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23973174 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23973188 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23973176 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23973184 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23973172 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23973185 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04600436 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23973181 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23973179 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04765302 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04600454 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23973162 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04937433 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23973165 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23973175 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23973177 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23973163 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10524033 - -
Affymetrix Mouse Gene 1.0 ST Array 10524023 - -
Affymetrix Mouse Gene 1.0 ST Array 10524029 - -
Affymetrix Mouse Gene 1.0 ST Array 10524032 - -
Affymetrix Mouse Gene 1.0 ST Array 10524025 - -
Affymetrix Mouse Gene 1.0 ST Array 10524021 - -
Affymetrix Mouse Gene 1.0 ST Array 10524028 - -
Affymetrix Mouse Gene 1.0 ST Array 10524031 - -
Affymetrix Mouse Gene 1.0 ST Array 10524030 - -
Affymetrix Mouse Gene 1.0 ST Array 10524024 - -
Affymetrix Mouse Gene 1.0 ST Array 10524026 - -
Affymetrix Mouse Gene 1.0 ST Array 10524027 - -
Affymetrix HT MG-430 PM Array Plate 1452134_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10524020 - -
Mouse Genome 430 2.0 Array 1452134_PM_at - -
Mouse Genome 430 2.0 Array 1452134_at - -
Mouse Genome 430 2.0 Array 1435109_at - -
Mouse Genome 430 2.0 Array 1435109_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 72392_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 72392_PM_at - -
Affymetrix Mouse Clariom S Array PSR0500009471.mm.1 TC0500001110.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500009471.mm.1 NM_001163531.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009471.mm.1 NM_001163531.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009471.mm.1 NM_001163532.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009471.mm.1 NM_001163532.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009471.mm.1 NM_028223.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009471.mm.1 NM_028223.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500009474.mm.1 TC0500001110.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500009474.mm.1 NM_001163531.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009474.mm.1 NM_001163531.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009474.mm.1 NM_001163532.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009474.mm.1 NM_001163532.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009474.mm.1 NM_028223.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009474.mm.1 NM_028223.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500009472.mm.1 TC0500001110.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500009472.mm.1 NM_001163531.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009472.mm.1 NM_001163531.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009472.mm.1 NM_001163532.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009472.mm.1 NM_001163532.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009472.mm.1 NM_028223.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009472.mm.1 NM_028223.3 Refseq
Mouse Expression Set 430 1452134_at - -
Mouse Expression Set 430 1452134_PM_at - -
Mouse Expression Set 430 1435109_at - -
Mouse Expression Set 430 1435109_PM_at - -
Mouse Genome 430A 2.0 Array 1452134_at - -
Mouse Genome 430A 2.0 Array 1452134_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1452134_at - -
Affymetrix HT Mouse Genome 430A Array 1452134_PM_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 102940278 - -
Illumina mouse-6 v1.0 expression beadchip 101770019 - -
Illumina mouse-6 v1.0 expression beadchip 3850390 - -
Illumina mouseRef-8 v1.0 expression beadchip 0003850390 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2677197 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2646438 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1236468 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1215917 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2677197 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2646438 - -
mouse-6 v1.1 (Illumina) scl0072392.2_133-S - -
Illumina Sentrix mouse-6.v1 bead array 3850390 - -
Illumina Sentrix mouse-6.v1 bead array 102940278 - -
Illumina Sentrix mouse-6.v1 bead array 101770019 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0072392.2_133-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 3850390 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 102940278 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 101770019 - -
MouseRef-8 v2 Expression BeadChip ILMN_2677197 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1236468 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2677197 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215917 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0