This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00512442_m1
See other CD200R3 GE Assays ›
Gene Symbol:
Cd200r3
Gene Name:
CD200 receptor 3
Gene Aliases:
4733401I18Rik, 4833409J19Rik, AI505817, BB106877, mCD200RLb
Chromosome Location:
Chr.16: 44943596 - 45058365 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00512442_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd200r3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001128132.1 NP_001121604.1 2 - 3 275 65
NM_001128133.1 NP_001121605.1 2 - 3 275 65
NM_027578.1 NP_081854.1 2 - 3 275 65
NM_029018.4 NP_083294.2 2 - 3 275 65
GenBank mRNA AK014637.1 - 2 - 3 249 65
AK014671.1 - 2 - 3 212 65
AK040868.1 - 2 - 3 262 65
AK170363.1 - 2 - 3 205 65
AY230199.1 - 2 - 3 249 65
AY703837.1 - 2 - 3 180 65
AY703838.1 - 2 - 3 180 65
AY703839.1 - 2 - 3 180 65
AY703840.1 - 2 - 3 180 65
AY703841.1 - 2 - 3 180 65
AY703842.1 - 2 - 3 180 65
BC106838.2 - 2 - 3 221 40044906 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000666.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cd200r3

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Process(es)

receptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600005846.mm.1 NM_001128132.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005846.mm.1 NM_001128132.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005846.mm.1 NM_001128133.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005846.mm.1 NM_001128133.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005846.mm.1 NM_027578.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005846.mm.1 NM_027578.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005846.mm.1 NM_029018.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005846.mm.1 NM_029018.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600005846.mm.1 TC1600000666.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline