This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00512837_g1
See other GPSM2 GE Assays ›
Gene Symbol:
Gpsm2
Gene Name:
G-protein signalling modulator 2 (AGS3-like, C. elegans)
Gene Aliases:
6230410J09Rik, LGN, Pins
Chromosome Location:
Chr.3: 108678638 - 108722330 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00512837_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpsm2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029522.2 NP_083798.2 6 - 7 1189 71
GenBank mRNA AK040996.1 - 6 - 7 1078 71
AK143552.1 - 6 - 7 1190 71
AY081187.1 - 6 - 7 1033 71
BC021308.1 - 5 - 6 744 5007832 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002750.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpsm2

Gene Ontology Categories:

Function(s) Process(es)

establishment of mitotic spindle orientation
lung epithelial cell differentiation
protein binding
GTPase regulator activity
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300021636.mm.1 NM_029522.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021636.mm.1 NM_029522.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300021636.mm.1 TC0300002750.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300021635.mm.1 NM_029522.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021635.mm.1 NM_029522.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300021635.mm.1 TC0300002750.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline