This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00512880_m1
See other TGDS GE Assays ›
Gene Symbol:
Tgds
Gene Name:
TDP-glucose 4,6-dehydratase
Gene Aliases:
2610017J16Rik, 2610025M23Rik, AI648925
Chromosome Location:
Chr.14: 118111911 - 118132765 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00512880_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tgds
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029578.3 NP_083854.3 3 - 4 256 86
GenBank mRNA AK011555.1 - 3 - 4 247 86
AK154341.1 - 3 - 4 233 86
AK165955.1 - 3 - 4 240 86
BC021419.1 - 3 - 4 232 5012877 86
BY710811.1 - 3 - 4 244 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002711.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tgds

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
nucleotide-sugar metabolic process
molecular_function
catalytic activity
dTDP-glucose 4,6-dehydratase activity
lyase activity
coenzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400020117.mm.1 NM_029578.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400020117.mm.1 NM_029578.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400020117.mm.1 TC1400002711.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400020116.mm.1 NM_029578.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400020116.mm.1 NM_029578.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400020116.mm.1 TC1400002711.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0