This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00512946_g1
See other LMF1 GE Assays ›
Gene Symbol:
Lmf1
Gene Name:
lipase maturation factor 1
Gene Aliases:
2400010G15Rik, AW822050, Tmem112, cld
Chromosome Location:
Chr.17: 25579174 - 25662826 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00512946_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lmf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029624.4 NP_083900.3 9 - 10 1432 66
NR_045520.1 7 - 8 1111 66
GenBank mRNA AK010312.1 - 9 - 10 1403 66
AK041659.1 - 7 - 8 1099 66
AK154622.1 - 9 - 10 1429 66
AK169983.1 - 9 - 10 1430 66
BC020104.1 - 9 - 10 1439 3963699 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000380.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lmf1

Gene Ontology Categories:

Function(s) Process(es)

ER to Golgi vesicle-mediated transport
protein secretion
protein glycosylation in Golgi
chylomicron remnant clearance
positive regulation of lipoprotein lipase activity
protein maturation
regulation of cholesterol metabolic process
regulation of triglyceride metabolic process
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700003172.mm.1 NM_029624.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700003172.mm.1 NM_029624.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700003172.mm.1 NR_045520.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700003172.mm.1 NR_045520.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700003172.mm.1 TC1700000380.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0