This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00513215_g1
See other MBOAT7 GE Assays ›
Gene Symbol:
Mboat7
Gene Name:
membrane bound O-acyltransferase domain containing 7
Gene Aliases:
5730589L02Rik, BB1, Leng4, Lpiat, Lpiat1, mBB1
Chromosome Location:
Chr.7: 3677789 - 3693525 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00513215_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mboat7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029934.3 NP_084210.2 2 - 3 867 99
GenBank mRNA AB055410.1 - 2 - 3 865 99
AK019981.1 - 2 - 3 278 99
AK038998.1 - 2 - 3 289 99
AK044149.1 - 2 - 3 294 99
AK150010.1 - 2 - 3 261 99
AK154606.1 - 2 - 3 866 99
AK158998.1 - 2 - 3 205 99
AK163428.1 - 2 - 3 281 99
AK170124.1 - 2 - 3 344 99
BC023417.1 - 2 - 3 315 5028759 99
BC118958.2 - 2 - 3 512 40093821 99
EU016380.1 - 1 - 2 79 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002135.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mboat7

Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
phospholipid biosynthetic process
ventricular system development
layer formation in cerebral cortex
phosphatidylinositol metabolic process
dihydrolipoamide branched chain acyltransferase activity
sterol O-acyltransferase activity
N-acetyltransferase activity
O-acyltransferase activity
palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
carnitine O-acyltransferase activity
acetyltransferase activity
C-acyltransferase activity
palmitoyltransferase activity
N-acyltransferase activity
acylglycerol O-acyltransferase activity
serine O-acyltransferase activity
O-acetyltransferase activity
O-octanoyltransferase activity
octanoyltransferase activity
O-palmitoyltransferase activity
S-acyltransferase activity
S-acetyltransferase activity
S-malonyltransferase activity
malonyltransferase activity
C-palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
succinyltransferase activity
N-succinyltransferase activity
O-succinyltransferase activity
S-succinyltransferase activity
sinapoyltransferase activity
O-sinapoyltransferase activity
peptidyl-lysine N6-myristoyltransferase activity
peptidyl-lysine N6-palmitoyltransferase activity
benzoyl acetate-CoA thiolase activity
3-hydroxybutyryl-CoA thiolase activity
3-ketopimelyl-CoA thiolase activity
N-palmitoyltransferase activity
myristoyltransferase activity
acyl-CoA N-acyltransferase activity
protein-cysteine S-myristoyltransferase activity
protein-cysteine S-acyltransferase activity
dihydrolipoamide S-acyltransferase activity
glucosaminyl-phosphotidylinositol O-acyltransferase activity
ergosterol O-acyltransferase activity
lanosterol O-acyltransferase activity
naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
2-methylhexanoyl-CoA C-acetyltransferase activity
butyryl-CoA 2-C-propionyltransferase activity
2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
L-2-aminoadipate N-acetyltransferase activity
UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
keto acid formate lyase activity
Ras palmitoyltransferase activity
azetidine-2-carboxylic acid acetyltransferase activity
peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
lysophospholipid acyltransferase activity
acetyl-CoA:L-lysine N6-acetyltransferase

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700019278.mm.1 TC0700002135.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700019278.mm.1 NM_029934.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700019278.mm.1 NM_029934.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700019287.mm.1 TC0700002135.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700019287.mm.1 NM_029934.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700019287.mm.1 NM_029934.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700019283.mm.1 TC0700002135.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700019283.mm.1 NM_029934.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700019283.mm.1 NM_029934.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700019280.mm.1 TC0700002135.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700019280.mm.1 NM_029934.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700019280.mm.1 NM_029934.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700019289.mm.1 TC0700002135.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700019289.mm.1 NM_029934.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700019289.mm.1 NM_029934.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700019288.mm.1 TC0700002135.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700019288.mm.1 NM_029934.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700019288.mm.1 NM_029934.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0