This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00513482_m1
See other POLR3H GE Assays ›
Gene Symbol:
Polr3h
Gene Name:
polymerase (RNA) III (DNA directed) polypeptide H
Gene Aliases:
5031409G22Rik, RPC8
Chromosome Location:
Chr.15: 81915030 - 81926247 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00513482_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr3h
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030229.4 NP_084505.2 1 - 2 429 87
GenBank mRNA AK019868.2 - 1 - 2 430 87
AK152517.1 - 1 - 2 402 87
BC010793.1 - 1 - 2 432 4221321 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001933.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polr3h

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

immune system process
nucleobase-containing compound metabolic process
transcription, DNA-templated
transcription from RNA polymerase III promoter
transcription initiation from RNA polymerase III promoter
innate immune response
defense response to virus
RNA polymerase III activity
DNA binding
DNA-directed RNA polymerase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500015187.mm.1 NM_030229.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500015187.mm.1 NM_030229.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500015187.mm.1 TC1500001933.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500015190.mm.1 NM_030229.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500015190.mm.1 NM_030229.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500015190.mm.1 TC1500001933.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500015189.mm.1 NM_030229.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500015189.mm.1 NM_030229.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500015189.mm.1 TC1500001933.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0