This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00513813_m1
See other BRE GE Assays ›
Gene Symbol:
Bre
Gene Name:
brain and reproductive organ-expressed protein
Gene Aliases:
6030405P19Rik, AI429776, B830038C02Rik
Chromosome Location:
Chr.5: 31697945 - 32084948 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00513813_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bre
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144541.1 NP_653124.1 4 - 5 354 78
NM_181279.1 NP_851796.1 6 - 7 465 78
NM_181280.1 NP_851797.1 5 - 6 363 78
NM_181281.1 NP_851798.1 3 - 4 198 78
NM_181282.1 NP_851799.1 4 - 5 275 78
GenBank mRNA AF440752.1 - 4 - 5 354 78
AF527952.1 - 6 - 7 465 78
AF527953.1 - 4 - 5 275 78
AF527954.1 - 4 - 5 285 78
AF527955.1 - 3 - 4 198 78
AF538925.1 - 5 - 6 363 78
AK050695.1 - 4 - 5 399 78
AK080991.1 - 4 - 5 390 78
AK156929.1 - 4 - 5 446 78
AK166884.1 - 4 - 5 390 78
AK197046.1 - 1 - 2 106 78
BC029277.1 - 4 - 5 705 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003709.mm.1

Additional Information:

This assay is in the 5'UTR region of the AF527953.1,AF527954.1,NM_181282.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bre

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
double-strand break repair
apoptotic process
cellular response to DNA damage stimulus
response to ionizing radiation
chromatin modification
G2 DNA damage checkpoint
positive regulation of DNA repair
tumor necrosis factor receptor binding
polyubiquitin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500002993.mm.1 TC0500003709.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500002993.mm.1 NM_144541.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002993.mm.1 NM_144541.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002993.mm.1 NM_181279.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002993.mm.1 NM_181279.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002993.mm.1 NM_181280.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002993.mm.1 NM_181280.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002993.mm.1 NM_181281.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002993.mm.1 NM_181281.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002993.mm.1 NM_181282.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002993.mm.1 NM_181282.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002994.mm.1 TC0500003709.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500002994.mm.1 NM_144541.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002994.mm.1 NM_144541.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002994.mm.1 NM_181279.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002994.mm.1 NM_181279.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002994.mm.1 NM_181280.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002994.mm.1 NM_181280.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002994.mm.1 NM_181281.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002994.mm.1 NM_181281.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002994.mm.1 NM_181282.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002994.mm.1 NM_181282.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002984.mm.1 TC0500003709.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500002984.mm.1 NM_181279.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002984.mm.1 NM_181279.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002984.mm.1 NM_181280.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002984.mm.1 NM_181280.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002984.mm.1 NM_181281.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002984.mm.1 NM_181281.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002984.mm.1 NM_181282.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002984.mm.1 NM_181282.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002986.mm.1 TC0500003709.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500002986.mm.1 NM_181279.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002986.mm.1 NM_181279.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002986.mm.1 NM_181280.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002986.mm.1 NM_181280.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4d4pq:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0